MBL™ Nessler Cylinder
Actual product may vary
Fisherbrand
MBL™ Nessler Cylinder
Manufactured with borosilicate glass specially selected for freedom from color or visible defects. MBL™ Nessler Cylinders are used for colorimetric analysis such as APHA color. Color of a sample substance is visually compared to a standard solution.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS12413601100 mL50 mL
Catalog number FKS12413601
Price (MYR)
283.00
Pack
Capacity (Metric):
100 mL
Graduations:
50 mL
Price (MYR)
283.00
Pack
Manufactured with borosilicate glass specially selected for freedom from color or visible defects. MBL™ Nessler Cylinders are used for colorimetric analysis such as APHA color. Color of a sample substance is visually compared to a standard solution.
  • Cylinder base: optically plane for maximum clarity
  • Fixed nominal stroke volume: available in 50 and 100mL
  • Graduations: marked at 50 and 100mL only
  • Units: sold in matched pairs for uniformity in comparisons
Specifications
Base TypeFlat
ShapeRound
Tolerance±0.8 mL
MaterialBorosilicate Glass
Diameter (Metric) Outside34 mm
For Use With (Application)Color Comparison of Samples
Capacity (Metric)100 mL
Height (Metric)180 mm
Product TypeNessler Cylinder
Graduations50 mL
Unit SizePack