TST™ Bowie Dick™ Test Pack
Actual product may vary
Fisherbrand
TST™ Bowie Dick™ Test Pack
Test the steam penetration and air removal capability of a vacuum sterilizer with this test pack. The TST™ Bowie Dick Test Pack is a unique combination of advance Browne intelligent ink technology and the original Bowie Dick concept.
Have Questions?
Catalog NumberSterilization Method
FKS12390489Steam, Vacuum
Catalog number FKS12390489
Price (MYR)
2,062.00
Pack
Sterilization Method:
Steam, Vacuum
Price (MYR)
2,062.00
Pack
  • Single-use test pack
  • A successful test confirms that steam penetration into a test pack is rapid and even and that air and other non-condensable gases have been effectively removed
  • The chemical indicator sheet at the center of the pack shows a defined color change from yellow to dark blue/purple when exposed to a specific combination of time, temperature and steam
  • Conforms to ISO 11140-4 Class 2
Specifications
AutoclavableAutoclavable
Certifications/ComplianceISO 11140-4(3) Class 2
For Use With (Application)For large porous load sterilizers
For Use With (Equipment)Vacuum sterilizer
Product TypeTST Bowie Dick Test Pack
Temperature (Metric)121°C to 124°C
Features121 to 124˚C for 8 to 8.3 min., Conforms to ISO 11140-4(3) Class 2
Quantity20 Pk.
Sterilization IndicatorYes
Sterilization Time8 to 8.3 min.
Sterilization MethodSteam, Vacuum
Unit SizePack