▸
Education Supplies
▸
General Classroom Products
▸
Magnifiers
▸
Digital Colony Counter Optional Magnifier
Actual product may vary
Fisherbrand
Digital Colony Counter Optional Magnifier
Use this magnifier with CC-200 Colony Counters.
Have Questions?
Catalog Number
Magnification
FKS12088183
3x
Catalog number
FKS12088183
Price (MYR)
1,873.00
Each
Add to cart
Magnification:
3x
Price (MYR)
1,873.00
Each
Add to cart
Use this magnifier with Stuart™ SC6 Colony Counters.
1.7x and 3x magnifier for CC-200 Colony Counter.
Pressure sensitive counting.
Average count facility.
Bright white energy saving LED lighting.
With BioCote antimicrobial protection.
Audible count confirmation.
Choice of light or dark background.
Connectivity to printer or computer.
Specifications
For Use With (Equipment)
The Colony Counter Stuart SCPPLUS
Includes
BioCote Antimicrobial protection
Quantity
1 Ea.
Magnification
3x
Product Type
Lens, Magnifying