903 Protein Saver Cards
Actual product may vary
Fisherbrand
903 Protein Saver Cards
Save time and costs during sample collection, transport, and archiving. Whatman™ 903 Protein Saver Card (EU) fits in the Whatman™ foil barrier resealable bags for storage.
Have Questions?
Catalog NumberProduct Type
FKS11962089Base Scale
Catalog number FKS11962089
Price (MYR)
3,685.00
Pack
Product Type:
Base Scale
Price (MYR)
3,685.00
Pack
Save time and costs during sample collection, transport, and archiving. Whatman™ 903 Protein Saver Card (EU) fits in the Whatman™ foil barrier resealable bags for storage.
  • Ready-to-use cards and kits
  • Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity
  • The sample collection area of our 903 Proteinsaver Card contains five half-inch circles
  • Each circle holds 75-80 μl of sample
  • Wraparound cover has spaces for name and date of collection and is imprinted with the universal biohazard symbol
Specifications
Product TypeBase Scale
Certifications/ComplianceISO 9001
Unit SizePack