OdoClave™ Deodorant Autoclave Pads
Actual product may vary
Fisherbrand
OdoClave™ Deodorant Autoclave Pads
Neutralize autoclave odors with a pleasant fragrance
Have Questions?
Catalog NumberScentColor
FKS11956072CherryRed
Catalog number FKS11956072
Price (MYR)
1,614.00
Pack
Scent:
Cherry
Color:
Red
Price (MYR)
1,614.00
Pack
Neutralize autoclave odors with a pleasant fragrance
  • Heat-release deodorant works automatically at autoclave temperatures
  • Open pouch and place in bag prior to autoclaving
  • Three-year shelf life or longer if stored in a sealed container
  • 32mm (1.75 in.) square
  • Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads
Specifications
Product TypeAutoclave Capsule
Color-codedNo
FormLiquid
ScentCherry
SterilityNon-sterile
AutoclavableAutoclavable
ColorRed
FormatPad
For Use With (Application)Autoclave
Unit SizePack