KDS100 Legacy Syringe Pump
Actual product may vary
Fisherbrand
KDS100 Legacy Syringe Pump
Accommodates 10μL to 60mL glass or plastic syringes of any make. Fisherbrand™ KDS100 Legacy Syringe Pump is an infusion pump with a fast flow rate.
Have Questions?
Catalog NumberNumber of Channels
FKS119397471
Catalog number FKS11939747
Price (MYR)
25,720.00
Each
Number of Channels:
1
Price (MYR)
25,720.00
Each
  • Simple menu-driven setup to program syringe diameter, flowrate and dispense volume
  • Backlit display
  • Automatic shutoff
  • Settings can be reviewed and changed during operation
  • Last settings are stored in memory for repeatability
Specifications
Amperage0.06 A
Voltage200/240 V
Flow Rate0.1 μL/hr. for 10 μL syringe, 426 mL/hr. for 60 mL syringe
Linear Force (Metric)9 kg
Reproducibility±0.1%
Dimensions (L x W x H) Exterior15.25 x 23 x 13 cm
Electrical Requirements200/240 V 50/60 Hz
Display TypeBacklit LCD
Number of Channels1