PYREX™ Heavy-Duty Griffin Beakers
Actual product may vary
PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
PYREX™ Heavy-Duty Griffin Beakers
Pyrex
PYREX™ Heavy-Duty Griffin Beakers
For use under harsh conditions such as mechanized washing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS119115162000 mL
FKS12569995400 mL
FIS02-555-25K4000 mL
Catalog number FKS11911516
Price (MYR)
2,319.00
Pack
Capacity (Metric):
2000 mL
Price (MYR)
2,319.00
Pack
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Specifications
Capacity (English)67.62 oz.
Diameter (English) Outer5.15 in.
Diameter (Metric) Outer13.1 cm
Capacity (Metric)2000 mL
MaterialBorosilicate Glass
For Use With (Application)Provides The Best Mechanical Strength Under Harsh Conditions Such as Mechanized Washings
Graduation Range200 to 1800 mL
Height (English)7.08 in.
Height (Metric)18 cm
Increments200 mL
Label TypeExtra Large Marking
Quantity4 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleDual Graduated Metric Scale
StyleGriffin
Unit SizePack