Check Order Status
Rubber Tubing
Actual product may vary
Fisherbrand
Rubber Tubing
For transferring liquids, gases and dilute acids.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
Diameter (Metric) Outer
Thickness (Metric)
FKS11895512
10 mm
14 mm
2 mm
FKS11587562
6.5 mm
16.5 mm
5 mm
2 Options
Catalog number
FKS11895512
Price (MYR)
407.00
EA
Add to cart
Diameter (Metric) Inner:
10 mm
Diameter (Metric) Outer:
14 mm
Thickness (Metric):
2 mm
Price (MYR)
407.00
EA
Add to cart
Higher resilience than most other polymers
Temperature range: -40°C to 70°C
Light and flexible
Not suitable for use with petroleum oils and hydrocarbon solvents (see neoprene tubing)
Specifications
Diameter (Metric) Inner
10 mm
Material
Rubber
Diameter (Metric) Outer
14 mm
Overall Length (Metric)
10 m
Thickness (Metric)
2 mm
For Use With (Application)
For transferring liquids, gases and dilute acids
Flexibility
Flexible