Check Order Status
Kimble Chase™ Borosilicate Glass 3.3 Reusable Test Tubes
Actual product may vary
Fisherbrand
Kimble Chase™ Borosilicate Glass 3.3 Reusable Test Tubes
Borosilicate Glass 3.3 Reusable Test Tubes are available in various metric diameters and are DIN 12395 certified.
Have Questions?
Catalog Number
Diameter (Metric)
Length (Metric)
Design Type
FKS11893370
12 mm
75 mm
Round Bottom, Flanged Collar
Catalog number
FKS11893370
Price (MYR)
231.00
PK
Add to cart
Diameter (Metric):
12 mm
Length (Metric):
75 mm
Design Type:
Round Bottom, Flanged Collar
Price (MYR)
231.00
PK
Add to cart
Use Kimble™ Borosilicate Glass 3.3 Reusable Test Tubes in the laboratory. They are available in various metric diameters and are DIN 12395 certified.
Available with or without rims
Round bottom
Autoclavable
Specifications
Certifications/Compliance
DIN 12395
Design Type
Round Bottom, Flanged Collar
Description
Thickness: 0.9 to 1.0mm
Length (Metric)
75 mm
Material
Borosilicate Glass
Quantity
100 Pack
Rimmed
Yes
Autoclavable
Autoclavable
Diameter (Metric)
12 mm