Specifications
Product TypeGlove Box Holder
ColorWhite
Depth (Metric)108 mm
Dimensions (D x W x H) Interior108 x 140 x 209 mm
Width (Metric)140 mm
Height (Metric)209 mm
HoldsGloves
MaterialEpoxy-coated Steel
MountingWall Mount
No. of Compartments1
Product StyleTop Loading
Capacity1 Box
Quantity1 Ea.
Unit SizeEach