Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes, 10mL
Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes, 10mL
Actual product may vary
Fisherbrand
Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes, 10mL
Use these tubes for centrifugation up to 2980 RCF. KIMAX™ Conical-Bottom Glass Centrifuge Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety. Manufactured from 51 expansion, low extractable borosilicate glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDisposableMaterialLength (Metric)Closure Type
FKS11828462Single-useBorosilicate Glass114 mmNot Included
FIS05-569-2Single-useBorosilicate Glass114 mmNot Included
Catalog number FKS11828462
Price (MYR)
131.00
PK
Add to cart
Disposable:
Single-use
Material:
Borosilicate Glass
Length (Metric):
114 mm
Closure Type:
Not Included
Price (MYR)
131.00
PK
Add to cart
Use these tubes for centrifugation up to 2980 RCF. KIMAX™ Conical-Bottom Glass Centrifuge Tubes are plastic shrink-wrapped in modular trays for cleanliness and safety. Manufactured from 51 expansion, low extractable borosilicate glass.
  • With conical shaped bottom
  • Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
Specifications
CalibrationNone
Certifications/ComplianceUSP, ASTM E 438
Capacity (Metric)10 mL
DisposableSingle-use
MaterialBorosilicate Glass
Thread Size15-415
ShapeConical
Length (Metric)114 mm
Bottom ShapeConical
Closure TypeNot Included
Diameter (Metric)16 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF2980 x g
Unit SizePK