Check Order Status
PTFE Micro Magnetic Stir Bar
Actual product may vary
Fisherbrand
PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11808892
13 mm
3 mm
SGE001810
10 mm
3 mm
FKS12656316
20 mm
3 mm
FKS16245851
30 mm
3 mm
FKS11888952
7 mm
2 mm
5 Options
Catalog number
FKS11808892
Price (MYR)
0.00
PK
Add to cart
Length (Metric):
13 mm
Diameter (Metric):
3 mm
Price (MYR)
0.00
PK
Add to cart
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Supplied in reusable snap cap vials.
For stirring in test tubes, small size beakers, or other containers.
Autoclavable.
Specifications
Autoclavable
Autoclavable
Certifications/Compliance
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color
White
Description
PTFE encapsulated Alnico V magnet core
For Use With (Equipment)
Containers & Vessels / Stirrer Hot Plates
Coating
PTFE
Diameter (Metric)
3 mm
Length (Metric)
13 mm
Quantity
10/Pk.
Shape
Cylinder
Type
Stir Bar, Micro