AutoclavableAutoclavable
ColorAssorted
HoldsPCR Tubes
MaterialPolypropylene
Positions1
ShapeRectangular
StackableYes
SterilityNon-sterile
For Use With (Equipment)0.2 mL, 0.5 mL and 1.5 mL Tubes
Quantity5 Pk.
Length (English)8.1 in.
Width (English)5.2 in.
Height (English)2.1 in.
Weight (English)2.2 lb.
Product TypePCR Tube Rack
ShelvesNone
Height (Metric)5.3 cm
Width (Metric)13.2 cm
Unit SizePack