Beaker, Low Form, With Spout
Actual product may vary
Fisherbrand
Beaker, Low Form, With Spout
Due to its shape, the beaker is a universal product for a wide range of laboratory applications.
Have Questions?
Catalog NumberCapacity (Metric)
FKS11732222600 mL
Catalog number FKS11732222
Price (MYR)
1,259.00
Pack
Capacity (Metric):
600 mL
Price (MYR)
1,259.00
Pack
Beakers are cylindrical vessels with a straight side wall. Thanks to this shape, beakers are universally usable laboratory vessels and usually serve as cooking glasses, collecting vessels and for preparing, mixing and heating solutions. Easy filling is possible via the large vessel opening. Beakers are available in various shapes and sizes: high and low, with or without spout, with or without scale.
  • Spout for clean pouring
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
Specifications
Diameter (Metric) Outer90 mm
Capacity (Metric)600 mL
MaterialBorosilicate Glass (3.3)
For Use With (Application)Heating; Diluting solutions; Mixing several components; Measuring liquid amounts; Sizes 5 and 10 mL without Retrace Code and graduation and size 10000 mL non-DIN/ISO size
No. per Pack10/Pk.
AutoclavableAutoclavable
Certifications/ComplianceASTM E-438 Type I, Class A
Unit SizePack