Narrow Mouth Wash Bottles
Narrow Mouth Wash Bottles
Actual product may vary
Fisherbrand
Narrow Mouth Wash Bottles
Wash bottles offer different colors of polypropylene closures for quick, at-a-glance identification of contents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeColor
FKS117244461000 mL38 mmNatural
FIS19-806-6451000 mL38 mmNatural
Catalog number FKS11724446
Price (MYR)
1,405.00
PK
Add to cart
Capacity (Metric):
1000 mL
Closure Size:
38 mm
Color:
Natural
Price (MYR)
1,405.00
PK
Add to cart
  • Low-density polyethylene
  • Bulk quantities available by special order
Specifications
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorNatural
MaterialLDPE
SterilityNon-sterile
Closure ColorNatural
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
MouthNarrow, Threaded
ShapeCylindrical
Product TypeEconomy Wash Bottle
Chemical LabelBlank
Closure MaterialPolypropylene
Closure Size38 mm
Unit SizePK