Cardboard Cryo Boxes with Water-Repellant Standard Coating
Actual product may vary
Fisherbrand
Cardboard Cryo Boxes with Water-Repellant Standard Coating
Combine these boxes with different grid inserts to hold a wide range of tube sizes. Fisherbrand™ Cardboard Cryo Boxes with Water-Repellant Standard Coating are temperature resistant down to -80°C, ideal for ultra-low temperature storage.
Have Questions?
Catalog NumberColorHeight (Metric)
FKS11557623White50 mm
Catalog number FKS11557623
Price (MYR)
278.00
Each
Color:
White
Height (Metric):
50 mm
Price (MYR)
278.00
Each
  • Highest flexibility: Cryo Boxes can be combined with different grid inserts for tubes from 6 to 28.5mm in diameter
  • The Cryo Boxes can be used for larger vessels if the grid is removed
Specifications
ColorWhite
MaterialCardboard
TypeCryo Box
For Use With (Application)Tubes from dia. 6mm to dia. 28.5mm
Holds6 mm to 28.5 mm Diameter Tubes
Quantity10 Pack
Height (Metric)50 mm
Unit SizeEach