Raised Scale Graduated Beaker
Actual product may vary
Fisherbrand
Raised Scale Graduated Beaker
Use these polypropylene beakers for a variety of general laboratory applications. VITLAB™ Raised Scale Graduated Beakers feature an ergonomically designed handle and an easy to read, embossed scale. Spout is designed to function correctly and accurately; unwanted drips are practically eliminated.
Have Questions?
Catalog NumberCapacity (Metric)
FKS103324225000 mL
Catalog number FKS10332422
Price (MYR)
1,493.00
Pack
Capacity (Metric):
5000 mL
Price (MYR)
1,493.00
Pack
  • Highly transparent polypropylene beaker
  • To preserve markings, do not clean at temperatures exceeding 60°C
  • Autoclavable at 121°C (2 bar) according to DIN EN 285
Specifications
ColorTransparent
Diameter (Metric) Outer210 mm
Capacity (Metric)5000 mL
MaterialPolypropylene
Height (Metric)270 mm
Quantity6 Pk.
TypeBeaker
AutoclavableAutoclavable
GraduatedYes
Certifications/ComplianceDIN EN 285
ScaleRaised Scale
Unit SizePack