Eutech™ ECFC7252201B pH/Temperature Combination Electrode
Product Image
Fisherbrand
Eutech™ ECFC7252201B pH/Temperature Combination Electrode
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberConnector Types
FKS10284752BNC
Catalog number FKS10284752
Price (MYR)
1,371.00
EA
Add to cart
Connector Types:
BNC
Price (MYR)
1,371.00
EA
Add to cart
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. This general purpose, plastic-body, double junction gel-filled pH combination electrode is used for general measurements. ECFC7252201B is available with 1 m cable.
Features
  • Parameter: pH/temperature
  • Liquid Junction Type: Porous HDPE pin
  • Sealed
Specifications
DescriptionGeneral purpose plastic-body pH electrode. Available with 1 m cable
Connector TypesBNC
JunctionDouble
MaterialPlastic Body
pH Range0 to 13
Temperature (Metric) Operating0°C to 80°C
TypepH Electrode
For Use With (Equipment)All pH meters with BNC input connector
Internal ReferenceAg/AgCl
Temperature (Metric) Range0°C to 80°C
Length (Metric)90 mm
Length (Metric) Cable1 m
Diameter (Metric) Body12 mm
Unit SizeEA