Strip Caps for Rack Packed Tubes
Actual product may vary
Fisherbrand
Strip Caps for Rack Packed Tubes
Manufactured from Polypropylene. BRAND™ Strip Caps for Rack Packed Tubes are compatible with standard microplates and suitable for working with multi-channel pipettors.
Have Questions?
Catalog NumberQuantity
FKS10233251120 Pack
Catalog number FKS10233251
Price (MYR)
436.00
Pack
Quantity:
120 Pack
Price (MYR)
436.00
Pack
  • Polypropylene tubes and rack are autoclavable at 121°C for 20min (except polyethylene caps)
  • Compatible with standard microplates and suitable for working with multi-channel pipettors
  • Size of tubes: outer diameter 8.8mm, height 45mm Ideal for biological tests, such as PCR*, cell uptake studies, RIA and EIA.
Specifications
Certifications/ComplianceDIN EN 285
Product Type8-cap Strip
Diameter (Metric) Outer8.8 mm
AutoclavableNot Autoclavable
MaterialPolyethylene
Quantity120 Pack
SterilityNon-sterile
For Use With (Equipment)Rack Packed Microtubes
Unit SizePack