Fisherbrand
Stir Bar Box Set, Cylindrical
Provides general purpose stirring for a wide variety of applications. Fisherbrand™ Stir Bar Box Sets, Cylindrical are isostatically molded in pure, FDA-approved PTFE plastic.
Have Questions?
Catalog NumberLength (Metric)
FKS10226853195 mm
Catalog number FKS10226853
Price (MYR)
391.00
Each
Length (Metric):
195 mm
Price (MYR)
391.00
Each
  • Guaranteed inert and leak-free
  • Smooth, round profile
  • Individually polished to stay cleaner for longer
  • Each set contains two (2) each of the following sizes:
6 x 10mm
4.5 x 15mm
6 x 20mm
6 x 25mm
6 x 30mm
8 x 40mm
8 x 50mm
10 x 60mm
10 x 70mm
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Dimensions (L x W x H)195 x 95mm
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Height (Metric)30mm
Includes18 Cylindrical Stirrer Bars
Width (Metric)95mm
Length (Metric)195 mm
Quantity1/Ea.
Unit SizeEach