PTFE Stir Bar Retriever
Actual product may vary
Fisherbrand
PTFE Stir Bar Retriever
Use to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS10226159450 mm
FKS10579081250 mm
Catalog number FKS10226159
Price (MYR)
198.00
Each
Length (Metric):
450 mm
Price (MYR)
198.00
Each
  • Inert and leak-free
  • Molded in pure, FDA-approved PTFE
  • Alnico V magnets
  • Autoclavable
Specifications
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Application)Removal of Stirrer Bars from vessels
For Use With (Equipment)PTFE Stirrer Bars
Height (Metric)10mm
Diameter (Metric)10 mm
Length (Metric)450 mm
Quantity1/Ea.
Unit SizeEach