Nonmetallic Oval Type I Safety Cans
Actual product may vary
Fisherbrand
Nonmetallic Oval Type I Safety Cans
Constructed of high-density polyethylene for resistance against denting, puncturing, and chipping. The nonmetallic oval Type I safety can stores liquids that may corrode stainless-steel cans.
Have Questions?
Catalog NumberCapacity (English)Color
FKS101805621 gal.Red
Catalog number FKS10180562
Price (MYR)
1,767.00
Each
Capacity (English):
1 gal.
Color:
Red
Price (MYR)
1,767.00
Each
Constructed of high-density polyethylene for resistance against denting, puncturing, and chipping.
  • Use with tough acids and corrosives, including liquids that corrode stainless-steel or steel-coated safety cans
  • Pressure-relief mechanism prevents buildup of vapors
  • Self-closing cap and flame arrester work together to prevent ignition sources from entering, without restricting liquid flow in or out of the can
  • Stainless-steel hardware for superior chemical resistance
  • Convenient trigger-spout mechanism and built-in handle for ease in dispensing and carrying
Specifications
Certifications/ComplianceFM, NFPA, OSHA
Product TypeSafety Can
DescriptionDent, Puncture, Chip Resistant
Capacity (English)1 gal.
Capacity (Metric)4 L
ColorRed
MaterialPolyethylene
IncludesFlame arrester and Stainless Steel hardware, trigger spout
ShapeOval
Length (English)7.6 in.
Length (Metric)19.3 cm
Width (English)4.6 in.
Width (Metric)11.6 cm
Height (English)12.75 in.
Height (Metric)32.3 cm
Diameter (English) Opening1.45 in.
Diameter (Metric) Opening3.6 cm
Unit SizeEach