Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Tubes
▸
Tube Caps and Closures
▸
Phenolic GPI Screw Caps for Disposable Culture Tubes
Actual product may vary
Corning
Phenolic GPI Screw Caps for Disposable Culture Tubes
With white rubber liner
Have Questions?
Catalog Number
Closure Size
Height (Metric)
FKS10150403
15-415
16 mm
Catalog number
FKS10150403
Price (MYR)
5,052.00
Pack
Add to cart
Closure Size:
15-415
Height (Metric):
16 mm
Price (MYR)
5,052.00
Pack
Add to cart
Made of a phenolic compound. These caps have a white rubber liner.
Specifications
Closure Size
15-415
Liner Material
Rubber
Product Type
Screw Cap
Autoclavable
Autoclavable
Material
Phenolic
Color
Black
Height (Metric)
16 mm
No. per Pack
1000/Pk.
No. per Case
1000/Cs.
Tip Material
Rubber
For Use With (Equipment)
Culture Tubes