Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Polypropylene Stopper
Actual product may vary
Fisherbrand
Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Polypropylene Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper is available with narrow neck or wide neck in a variety of sizes. DE-M marking on the neck provides quick and accurate volume check. Polypropylene stopper included.
Have Questions?
Catalog NumberCapacity (Metric)Neck Style
FKS101205715 LNarrow
Catalog number FKS10120571
Price (MYR)
1,856.00
Each
Capacity (Metric):
5 L
Neck Style:
Narrow
Price (MYR)
1,856.00
Each
  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • DAkkS calibration certificate available upon request
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • DE-M marking provides quick and accurate volume check
  • Polypropylene stopper included
Specifications
CalibrationCalibrated To Contain
Certifications/ComplianceDIN EN ISO 1042
Closure MaterialPolypropylene
ColorClear
DescriptionDE-M Marking
Capacity (Metric)5 L
MaterialBorosilicate Glass
Limit of Error±1.2 mL
Neck StyleNarrow
No. per Case1 Each
GraduatedYes
ClassA
Closure TypeStopper
Closure Size34/35
Unit SizeEach