Wheaton™ Serum Bottles and Vials
Actual product may vary
Wheaton™ Serum Bottles and Vials
Wheaton™ Serum Bottles and Vials
Fisherbrand
Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Color
FKS10071382125 mLClear
FKS10021362100 mLClear
Catalog number FKS10071382
Price (MYR)
12,775.00
Pack
Capacity (Metric):
125 mL
Color:
Clear
Price (MYR)
12,775.00
Pack
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Specifications
Capacity4.23 oz., 125 mL
Capacity (Metric)125 mL
ColorClear
Diameter (English)2.12 in.
MaterialBorosilicate Glass
Height (Metric)107 mm
Height (English)4.21 in.
Diameter (Metric)54 mm
Capacity (English)4.23 oz.
Unit SizePack