▸
Salts and Inorganics
▸
Inorganic Salts
▸
Transition Metal Salts
▸
Iron(II) Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Iron(II) Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
250 g
1 kg
Catalog number
FKCI115753
also known as FKCI/1157/53
Price (MYR)
453.00
Bottle
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
453.00
Bottle
Add to cart
Specifications
Melting Point
64°C
CAS
7782-63-0
Color
Blue-Green
pH
3 to 4
Physical Form
Solid
View more