▸
Acids
▸
Inorganic Acids
▸
Boric Acid
▸
Boric Acid, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Boric Acid, Certified AR for Analysis, Fisher Chemical™
H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C | CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change view
Quantity:
1 kg
500 g
Catalog number
FKCB380060
also known as FKCB/3800/60
Price (MYR)
246.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
246.00
Each
Add to cart
Specifications
pH
3.8 to 4.8
Color
White
CAS
10043-35-3
Melting Point
169°C
Packaging
Plastic powder jar
View more
H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C