UV Blocking Eyewear
UV Blocking Eyewear
Actual product may vary
UVP
UV Blocking Eyewear
Designed to block ultraviolet radiation of 254 to 365nm as well as longwave blue haze. UVP™ UV Blocking Eyewear provides optimum contrast in viewing and lessened eye fatigue with spectacles, goggles and face shields that minimize eye and face UV exposure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLens MaterialProduct Type
FISUVP98000204PlasticUVC-803 Face Shield
FISUVP38015101PlasticReplacement Faceshield
Catalog number FISUVP98000204
Price (MYR)
1,435.00
EA
Add to cart
Lens Material:
Plastic
Product Type:
UVC-803 Face Shield
Price (MYR)
1,435.00
EA
Add to cart
  • Designed to block ultraviolet radiation of 254 to 365nm as well as longwave blue haze
  • Provide optimum contrast in viewing and lessened eye fatigue
UVC-303 Spectacles
  • Made from impact-resistant polycarbonate
  • Top and side-shield design
  • Dimensions: 51 × 152mm (2 × 6 in.)
UVC-503 Goggles
  • Made from specially formulated plastic, offering optimum contrast and lessened eye fatigue
  • Lightweight design and flexible rubber trip for comfortable wearing
  • Air vents
  • Dimensions: 76 × 210mm (3 × 6 in.)
UVC-803 face shield
  • Covers face and neck
  • Made from plastic
  • Dimensions: 432 × 210mm (17 × 8.25 in.)
  • For exposure periods longer than eight hours or for ultraviolet wavelength emis-
  • sions other than 254nm to 365nm, consult Document HSM 73-11009 Criteria for a Recommended Standard ... Occupational Exposure to Ultraviolet Radiation published by the US Department of Health, Education and Welfare.
  • Eyewear is designed to protect eyes, face and/or neck. Care must be taken to protect other parts of the body from severe skin burn associated with short-wave UV radiation. Avoid shining any ultraviolet lamp on reflective surfaces. Wear protective clothing and skin cream.
Specifications
Lens CoatingUV-treated
Certifications/ComplianceComplies with ANSI Z87.1; NIOSH
Lens TintClear
Product TypeUVC-803 Face Shield
For Use With (Equipment)UV blocking Face Shield
Lens MaterialPlastic
Unit SizeEA