KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Actual product may vary
Fisherbrand
KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm), for routine filtration of corrosive liquids and removal of particulates from HPLC solvents.
Have Questions?
Catalog NumberVolume (Metric)
FISK953825-0000300 mL (Funnel), 1000 mL (Flask)
Catalog number FISK953825-0000
Price (MYR)
5,599.00
Each
Volume (Metric):
300 mL (Funnel), 1000 mL (Flask)
Price (MYR)
5,599.00
Each
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
  • Support base has a 40-60 micron porosity glass frit and an integral vacuum connection located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
  • Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47mm fritted glass support base and a filtration flask
  • Joints are 40/35
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
DisposableReusable
MaterialGlass
SterilityNon-sterile
TypeMicrofiltration Assembly
Volume (Metric)300 mL (Funnel), 1000 mL (Flask)
For Use With (Equipment)Filtration Assembly and vacuum pump
Unit SizeEach