Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Actual product may vary
Fisherbrand
Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingSterility
FISK7496250030Single packSterile
FISK7496250010BulkNon-sterile
Catalog number FISK7496250030
Price (MYR)
1,982.00
CS
Add to cart
Packaging:
Single pack
Sterility:
Sterile
Price (MYR)
1,982.00
CS
Add to cart
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
  • Clear polypropylene tube
  • Polyacetal pestle with molded-in shaft guard
  • Abrasive surface (pestle tip and inner tube area)
  • Available in Sterile and non-sterile versions
  • Capacity: 1.5mL
For animal tissues and organs, plant materials, insects, DNA, RNA, proteins, yeasts and enzymes
Specifications
Capacity (English)0.05 oz.
For Use With (Application)For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes
AutoclavableAutoclavable
ColorClear
IncludesPP tube, POM pestle
Capacity (Metric)1.5 mL
PackagingSingle pack
Capacity0.05 oz., 1.5mL