Fisherbrand™ Rectangular TLC Developing Tank
Actual product may vary
Fisherbrand
Fisherbrand™ Rectangular TLC Developing Tank
Rectangular TLC Developing Tank is a glass 'Shorty' tank accommodating 5x10cm, 10x10cm, or 10x20cm TLC plates. Features a centering baffle for plate support during development and is supplied with a lid. Made from 90 expansion soda lime glass to USP Type III.
Have Questions?
Catalog NumberForUseWithEquipment
FISK416180-1020
Catalog number FISK416180-1020
Price (MYR)
1,925.00
Case
Price (MYR)
1,925.00
Case
  • Accommodates 5 x 10 cm, 10 x 10 cm, or 10 x 20 cm TLC plates turned long side down
  • Centering baffle to support plates during development
  • Supplied with lid
  • Beveled edges with tops and bottoms ground flat
  • Manufactured from 90 expansion soda lime glass conforming to USP Type III requirements
Specifications
Closure TypeStandard
MaterialSoda Lime Glass
Quantity1 Ea.
TypeRectangular TLC Developing Tank
Holds20 H x 10 mm W Plate
ShapeRectangular
Unit SizeCase