▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
Crucibles
▸
Porcelain Gooch Filtering Crucibles
Actual product may vary
Fisherbrand
Porcelain Gooch Filtering Crucibles
For total suspended solids determinations
Have Questions?
Catalog Number
Capacity (Metric)
Diameter (Metric) Outer
Height (Metric)
Thickness (Metric)
FISFB969A
25 mL
36mm
45mm
2mm
Catalog number
FISFB969A
Price (MYR)
384.00
Pack
Add to cart
Capacity (Metric):
25 mL
Diameter (Metric) Outer:
36mm
Height (Metric):
45mm
Thickness (Metric):
2mm
Price (MYR)
384.00
Pack
Add to cart
Glazed on the outside, except for bottom surface and rim
Perforated bottom
Withstand temperatures up to 1150°C (2102°F)
Without cover
Specifications
Material
Porcelain
For Use With (Application)
For total suspended solids determination
Max. Temperature (Metric)
1150°C
Thickness (Metric)
2mm
Capacity (Metric)
25 mL
Diameter (Metric) Outer
36mm
Height (Metric)
45mm
Product Type
Gooch Filtering Crucible