Solvent-Vented™ and Safety-Labeled Wash Bottles
Actual product may vary
Solvent-Vented™ and Safety-Labeled Wash Bottles
Solvent-Vented™ and Safety-Labeled Wash Bottles
Solvent-Vented™ and Safety-Labeled Wash Bottles
Fisherbrand
Solvent-Vented™ and Safety-Labeled Wash Bottles
Pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Label
FISFB0340937Acetone
FISFB0340936Ethanol
FISFB0340935Isopropanol
FISFB0340934Unlabeled
Catalog number FISFB0340937
Price (MYR)
-
Chemical Label:
Acetone
  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Specifications
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
MaterialLDPE
Closure ColorRed
Certifications/ComplianceOSHA 29 CFR 1910.1200
MouthWide
Chemical LabelAcetone
Closure MaterialPolypropylene
Closure Size53 mm