Wiretrol™ I Calibrated Micropipets
Actual product may vary
Fisherbrand
Wiretrol™ I Calibrated Micropipets
Easy-to-read volume line won't wash off
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FIS21-175F100 μL
FIS21-175B10 μL
5000-102020 μL
Catalog number FIS21-175F
Price (MYR)
-
Volume (Metric):
100 μL
  • Deliver within 1%
  • One-handed operation
  • Accepts both high- and low-viscosity liquids including corrosives
  • Color coded
  • Micropipet 21-175D, -175E and -175F have PTFE-fluorocarbon–resin tips on plunger
Specifications
CalibrationTo Contain
MaterialGlass
For Use With (Application)For a wide variety of applications, Wiretrol™ will accept both high and low viscosity liquids including corrosives
IncludesHandy dispenser cap, handle, Stainless Steel plunger
ColorBlue
Volume (Metric)100 μL
TypeCalibrated Micropipet
Unit SizePack