Pop-Up Drum Fill Gauge
Pop-Up Drum Fill Gauge
Actual product may vary
Justrite
Pop-Up Drum Fill Gauge
Prevents overfillin when dispensing waste into drums
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)
FIS19-05906711 in.
FIS17-986-555 in.
Catalog number FIS19-059067
Price (MYR)
233.00
Each
Add to cart
Length (English):
11 in.
Price (MYR)
233.00
Each
Add to cart
  • Features handy indicator that pops up when drum is almost full
  • Nonmetallic
  • Fits 0.75 in. (1.9cm) bung
  • Available in two lengths
Specifications
Length (Metric)27.94 cm
For Use With (Application)To prevent overfilling when dispensing waste into drums
Length (English)11 in.
MaterialPolyethylene
Product TypeDrum Fill Gauge
Unit SizeEach