PREempt™ RTU Disinfectant Solution
Actual product may vary
Corning
PREempt™ RTU Disinfectant Solution
Cleans and disinfects environmental surfaces in cleanrooms and laboratory areas including work stations, fume hoods, equipment and other hard non-porous environmental surfaces. Contec™ PREempt™ RTU Disinfectant Solution helps to ensure user, protocol and product compliance with fast contact times and broad-spectrum efficacy.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (English)Container Type
FIS19-039-93332 oz.Spray bottle
CTCSBT32CQ32 oz.Spray bottle
Catalog number FIS19-039-933
Price (MYR)
948.00
Case
Volume (English):
32 oz.
Container Type:
Spray bottle
Price (MYR)
948.00
Case
  • AHP - Utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients that when combined with low levels of hydrogen peroxide dramatically increase potency and cleaning performance
  • Cleaner - Effective disinfectant as well as a cleaner resulting in added confidence that disinfection can occur
  • Faster - Rapid and realistic contact times on a broad spectrum of difficult to kill pathogens
  • Responsible - Designed to be easier on surfaces, staff and occupants. Users are not exposed to VOCs (volatile organic compounds) and PPE is not required as per health and safety rating
  • 5 minute tuberculocidal
  • 10 minute fungicidal
  • 30 seconds broad-spectrum sanitizing
  • 1 minute bactericidal, virucidal
Specifications
Product TypeDisinfectant Solution
Container TypeSpray bottle
Volume (English)32 oz.
Volume (Metric)0.9 L
Unit SizeCase