Specifications
AntigenSLC35D2
ApplicationsImmunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
ClassificationPolyclonal
ConjugateUnconjugated
DilutionImmunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200
FormulationPBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No.Q76EJ3
Gene AliasHFRC, HFRC1UDP-N-acetylglucosamine transporter, Homolog of Fringe connection protein 1, MGC117215, MGC142139, Solute carrier family 35 member D2, solute carrier family 35, member D2, SQV7Lfringe connection, SQV7-like protein, UDP-galactose transporter-related protein 8, UDP-N-acetylglucosamine/UDP-glucose/GDP-mannose transporter, UGTREL8, UGTrel8hfrc
Gene SymbolsSLC35D2
Host SpeciesRabbit
ImmunogenThis antibody was developed against a recombinant protein corresponding to amino acids: YSFLTLSSQLKPKPVGEENICLD
Purification MethodAffinity Purified
Quantity0.1 mL
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)11046
Test SpecificitySpecificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target SpeciesHuman
IsotypeIgG
Unit SizeEach