Long Blade Scissors
Actual product may vary
Excelta
Long Blade Scissors
For long cuts
Have Questions?
Catalog NumberLength (Metric)
FIS17-467-48412.7 cm
Catalog number FIS17-467-484
Price (MYR)
113.00
Each
Length (Metric):
12.7 cm
Price (MYR)
113.00
Each
  • Fine precision
  • Stainless steel construction
Specifications
Length (English)5 in.
Length (Metric)12.7 cm