AutoclavableNot Autoclavable
ColorRed
HoldsFlasks (Assorted Sizes)
MaterialVinyl-coated Lead
Positions1
ShapeCylindrical
StackableNo
SterilityNon-sterile
For Use With (Equipment)Flasks
Length (Metric)30 cm
Diameter (English) Outer0.25 in.
Diameter (Metric) Outer0.64 cm
Weight (English)0.25 lb.
Weight (Metric)114 g
Product TypeLead Ring Flask Weight
Diameter (Metric) Inner20 mm
ShelvesNone
Height (Metric)20 mm
Width (Metric)20 mm
Unit SizeEach