Specifications
Dimensions (L x W x H)30 x 20 x 2 in.
Height (Metric)5.08 cm
Length (Metric)76.2 cm
MaterialFiberglass
TypeReagent Tray
Width (Metric)51 cm
For Use With (Application)Lightweight tray resists Acids, mild alkalis and most solvents
Length (English)30 in.
Height (English)2 in.
ShapeRectangular
Width (English)20 in.
Unit SizeEach