Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Actual product may vary
Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Fisherbrand
Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)Temperature Range (English)AccuracyModel
FIS14-648-44-50°C to +300°C-58°F to +572°F±1°CShockproof/Water-resistant Thermometer
CTL14-648-44-50°C to +300°C-58°F to +572°F±1°CShockproof/Water-resistant Thermometer
CTL06-664-27-50°C to +300°C-58°F to +572°F±0.4°CUltra Shockproof/Water-resistant Thermometer
Catalog number FIS14-648-44
Price (MYR)
300.00
Each
Temperature Range (Metric):
-50°C to +300°C
Temperature Range (English):
-58°F to +572°F
Accuracy:
±1°C
Model:
Shockproof/Water-resistant Thermometer
Price (MYR)
300.00
Each
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
  • Short-stem piercing models are ideal for carrying in a pocket
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
  • Traceable to standards provided by NIST
  • Waterproof thermometers (see Models 14-648-43, -44) are ideal for wet labs, wash-down areas and field use
  • 0.375 in. LCD screen thermometers available (see Cat. No.14-648-47); also available with protective guards (see Cat. Nos. 06-664-32 and 06-664-30)
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Depth (English)0.5 in.
Depth (Metric)1.27 cm
Diameter (English) Probe0.14 in.
For Use With (Application)Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
IncludesStainless steel probe, Probe cover and Silver oxide battery
Length (English) Probe8 in.
Length (Metric)260 mm
Length (Metric) Probe20 cm
MaterialStainless Steel
ModelShockproof/Water-resistant Thermometer
Resolution0.1°C
Weight (English)0.87 oz.
Weight (Metric)24.6 g
Width (English)1.75 in.
Product TypeDigital Thermometer
Length (English)10.25 in.
Max. Temperature (English)572°F
Max. Temperature (Metric)300°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +572°F
Temperature Range (Metric)-50°C to +300°C
CalibratedYes
Battery Type1.5V Silver Oxide, 357BP
Accuracy±1°C
DisplayDigital LCD
Width (Metric)4 cm
Unit SizeEach