PYREX™ Reusable Class A Volumetric Pipets
Actual product may vary
Corning
PYREX™ Reusable Class A Volumetric Pipets
Borosilicate-glass with fire-polished top and tip manufactured and calibrated To Contain(TC) and To Deliver(TD)
Have Questions?
Catalog NumberVolume (Metric)ToleranceTypeColor
FIS13-646-321 mL±0.01 mLVolumetric PipetBlue
Catalog number FIS13-646-32
Price (MYR)
963.00
Case
Volume (Metric):
1 mL
Tolerance:
±0.01 mL
Type:
Volumetric Pipet
Color:
Blue
Price (MYR)
963.00
Case
Calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969. Each pipet is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. These precision pipets are manufactured and calibrated “To Contain” (bottom line) and “To Deliver” (top line) to specifications for Class A volumetric ware. Sizes 1 mL through 25 mL are color-coded with colored graduations enameled onto the glass.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Tested and calibrated in an ISO/IEC 17025 certified laboratory.
Specifications
AutoclavableAutoclavable
Certifications/ComplianceASTM E542, ASTM E969
ClassA
MaterialBorosilicate Glass
SterilityNon-sterile
Length (English)12.32 in.
Tip StyleRound
Tolerance±0.01 mL
Volume (English)0.033 oz.
ColorBlue
Volume (Metric)1 mL
Length (Metric)31.3 cm
TypeVolumetric Pipet
DisposableReusable
For Use With (Equipment)Mechanical Pipetting Device
Unit SizeCase