Microslide Cabinets, One drawer, holds 100 slide holders/400 slides
Actual product may vary
Fisherbrand
Microslide Cabinets, One drawer, holds 100 slide holders/400 slides
Store microscope slides with ease, speed, efficiency
Have Questions?
Catalog number FIS12-588-30
Price (MYR)
-
Store microscope slides with ease, speed, efficiency
  • High-strength steel drawers
  • Each drawer holds up to 100 four-place microscope slide holders (available separately, Cat. No. 12-588-25)
  • Includes inside door stop to prevent spills, lock compressor to keep slide holders in place
  • Gray enamel finish
  • Have interlocking mating slots for stacking for modular storage system
  • Equipped with rubber feet for placement on lab bench
Specifications
TypeMicro Slide Storage Cabinet
Slide Capacity100 slide holders/400 slides
ColorGray Enamel (Finish)
Count (Drawers)1
Depth (English)16.25 in.
Depth (Metric)419 mm
Device Holds400 Slides
Height (English)5.12 in.
Height (Metric)130.04 mm
IncludesInside door stop to prevent spills, lock compressor to keep slide holders in place
MaterialSteel
Width (English)6.5 in.
Width (Metric)165.1 mm
Unit SizeEach