▸
Microscope Slides
▸
Glass Microscope Slides
▸
Economy Plain Glass Microscope Slides
Actual product may vary
Fisherbrand
Economy Plain Glass Microscope Slides
Cost-effective slide for general-purpose applications
Have Questions?
Catalog number
FIS12-550-A3
Price (MYR)
155.00
Pack
Add to cart
Price (MYR)
155.00
Pack
Add to cart
Cost-effective slide for general-purpose applications
Plain white glass slide
Precision ground edges on all four sides
Corrosion resistant
ThicknessL 1mm
Specifications
Edges
Ground
Length (Metric)
25 mm
Material
Glass
Thickness (Metric)
1 mm
Type
Microscope Slide
Width (Metric)
75 mm
Color
White