Search Fisher Scientific
| Catalog Number | Capacity (Metric) |
|---|---|
| FIS10-210-5G | 1 L |
| FIS10-210-5C | 100 mL |
| FIS10-210-5E | 250 mL |
| FIS10-210-5A | 25 mL |
The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-beaded, heavy-tubing necks, tooled for polyethylene stopper. The stopper is made with a closed bottom and is of linear high density polyethylene to conform to standard taper stopper dimensions. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. The 2 mL size is test tube shaped.