Nalgene™ Polypropylene Graduated Cylinders
Actual product may vary
Nalgene™ Polypropylene Graduated Cylinders
Nalgene™ Polypropylene Graduated Cylinders
Fisherbrand
Nalgene™ Polypropylene Graduated Cylinders
Conduct applications accurately with these graduated cylinders, which feature generous pour spouts and molded-in graduations.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FIS08-572H2000 mL20 mL
Catalog number FIS08-572H
Price (MYR)
958.00
Each
Capacity (Metric):
2000 mL
Graduations:
20 mL
Price (MYR)
958.00
Each
Conduct applications accurately with Thermo Scientific™ Nalgene™ Polypropylene Graduated Cylinders, which feature generous pour spouts, molded-in graduations and a large blue base. There is no meniscus to confuse readings, eliminating guesswork; the cylinders can be chemically sterilized and are suitable for food and beverage use. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
  • Excellent chemical resistance for a wide variety of solutions
  • Resists temperatures up to 135°C (275°F) for a variety of uses
  • Autoclavable (autoclaving will affect accuracy)
  • Molded-in graduations are silk-screened for easy reading (10mL to 1L sizes only)
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Ordering Information:
Individually packaged.
Recycling Information: Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene labware at end of product life in communities where polypropylene can be recycled.
Nalgene beakers and graduated cylinders are delivered in corrugate cartons made from 100% post-consumer recycled material.  Packaging bags are made from LDPE and are readily recyclable once the label is removed.   Please continue the cycle and recycle your cartons and bags.
.
Specifications
Capacity (Metric)2000 mL
MaterialPolypropylene
Tolerance±12 mL
Certifications/ComplianceASTM E1272, CFR 21, ISO 6706, Part 177.1520
CalibrationTo Contain, To Deliver
Diameter (English)4.08 in.
Scale200mL to 2000mL in 20mL increments
Diameter (Metric)103.6 mm
Quantity1/Pk., 4/Cs.
Graduations20 mL
AutoclavableAutoclavable
Limit of Error±12.0mL
ShapeRound
Max. Temperature (English)250°F
Base TypeHexagonal
Height (English)20.6 in.
Height (Metric)523.2 mm
ACT LabelYes
Product TypeGraduated Cylinder
Max. Temperature (Metric)121°C
Diameter (English) Inner3.19 in.
Diameter (Metric) Inner81 mm
Unit SizeEach