Plasticoid Walter Crucible Holder
Actual product may vary
Fisherbrand
Plasticoid Walter Crucible Holder
Rubber with removable glass stem ground at 45° angle
Have Questions?
Catalog number FIS08-285
Price (MYR)
102.00
Each
Price (MYR)
102.00
Each
Description
  • Holds Gooch Crucible
  • Fits neck of filter flasks that take rubber stopper sizes 6, 7, or 8
  • The top of the glass stem, inside the holder, is flared to collect filtrate
  • With glass stem, but without filter flask or crucibles
Order Info
  • Tapered rubber portion, 1.5 in. (38mm) at top and 0.875 in. (22mm) at the bottom.
Specifications
ColorBlack
MaterialRubber
For Use With (Application)Used with Suction Flask
TypePlasticoid Walter Crucible Holder
Height (Metric)60 mm
Unit SizeEach