▸
Clamps and Supports
▸
Clamps
▸
Polypropylene Clamp with Flow Control
Actual product may vary
Fisherbrand
Polypropylene Clamp with Flow Control
Regulates flow through tubing from fully open to closed.
Have Questions?
Catalog Number
Color
FIS05-869
White
Catalog number
FIS05-869
Price (MYR)
35.00
Pack
Add to cart
Color:
White
Price (MYR)
35.00
Pack
Add to cart
Fits 0.12 to 0.5 in. (3.2 to 13mm) O.D. tubing
Molded white polypropylene construction
Autoclavable
With 15-position control
Will not distort, rust, or corrode
Specifications
Autoclavable
Autoclavable
Type
Tubing Clamp
Material
Polypropylene
For Use With (Equipment)
Tube with 0.12 to 0.5 in. (0.304 to 1.27cm) OD
Length (English)
1.5 in.
Length (Metric)
3.81 cm
Size
Medium
Color
White