STEMdiff™ Neural Rosette Selection Reagent
Actual product may vary
Fisherbrand
STEMdiff™ Neural Rosette Selection Reagent
Enzyme-free reagent for the selective detachment of neural rosettes
Have Questions?
Catalog NumberQuantity
FIS05-832100 mL
Catalog number FIS05-832
Price (MYR)
203.00
Pack
Quantity:
100 mL
Price (MYR)
203.00
Pack
  • ​STEMdiff™ Neural Rosette Selection Reagent is an enzyme-free reagent for the selective detachment of neural rosette clusters from adherent neural aggregates previously generated from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells using STEMdiff™ Neural Induction Medium, without manual scraping
  • Collecting and re-plating rosette clusters after incubation with STEMdiff™ Neural Rosette Selection Reagent will yield highly pure populations of neural progenitor cells, which can be further sub-cultured as single cells
Specifications
Cell TypeNeural Cells, PSC-Derived, Pluripotent Stem Cells
Quantity100 mL
Unit SizePack