I-Chem™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
Actual product may vary
I-Chem™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
I-Chem™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
I-Chem™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
I-Chem™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
Thermo Scientific
I-Chem™ Boston Round Narrow-Mouth Amber Glass Bottles with Closure
Select these round, narrow-mouth, amber glass jars with closure for collecting water samples for a variety of organic and inorganic target analytes where light protection is required.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)LevelClosure Size
FIS05-719-183250 mLCertified24-414 mm
FIS05-719-78500 mLCertified28-400 mm
FIS05-719-911000 mLCertified33-430 mm
Catalog number FIS05-719-183
Price (MYR)
453.00
Case
Capacity (Metric):
250 mL
Level:
Certified
Closure Size:
24-414 mm
Price (MYR)
453.00
Case
Designed for collecting water samples for a variety of organic and inorganic target analytes where light protection is required. Thermo Scientific™ Boston Round Narrow-Mouth Amber Glass Bottles are available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses, Processed the same way as the certified bottles but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
  • Amber Type III soda-lime glass bottle; recommended for light sensitive samples
  • PTFE-lined white polypropylene closed-top cap
  • Sizes range from 125 to 1000mL
  • Closures assembled on bottles
Includes:
Certified bottles come with the following features:
  • Certified to meet or exceed EPA performance-based standards for semi-volatile organics, pesticides, PCBs and metals analyses
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each bottle individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping
Processed bottles are processed in the same manner as certified bottles and come with all the features of certified bottles, but bottles are not barcoded, and CofA is not included.
Unprocessed bottles are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; overpack required for shipping.
Specifications
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Certification ParametersSemi-VOA, pesticides, PCBs
ColorAmber
DescriptionAmber Glass Boston Round
Diameter (English) Inner0.63 in.
Diameter (English)2.36 in.
Diameter (Metric) Inner1.6 cm
MaterialGlass (Type 3)
No. per Case12
Replacement ClosureB267-2414UBB
Closure ColorWhite
LevelCertified
Liner MaterialPTFE
Volume (English) Brim8.5 oz.
Volume (Metric) Brim250 mL
MouthNarrow
Height (Metric)136 mm
Height (English)5.35 in.
Closure MaterialPolypropylene
Closure Size24-414 mm
Diameter (Metric)60 mm
Closure TypeClosed-top Cap
Capacity (English)8 oz.
Certifications/ComplianceCertified and Processed bottles meet or exceed EPA performance based standards for semi-volatile organics, pesticides, and PCB's analysis; manufactured under an ISO certified quality management system. Certificate of Analysis (CofA) included.
Product TypeSample Bottle
Unit SizeCase